Learn More
LICORbio NewBlot™ Nitro Stripping Buffer for Nitrocellulose Membranes (100 mL)
Catalog No. NC9741227
Encompass_Preferred
NewBlot Nitro is optimized for near-infrared (NIR) Western blots and is specially formulated for use with IRDye secondary antibodies, including IRDye 800CW, IRDye 680RD, and IRDye 680LT. NewBlot Nitro is intended for nitrocellulose membranes only, and is ideal for removal of secondary antibody.
Catalog No. NC9741227 Supplier LICORbio Supplier No. 92840030
NewBlot Nitro is optimized for near-infrared (NIR) Western blots and is specially formulated for use with IRDye secondary antibodies, including IRDye 800CW, IRDye 680RD, and IRDye 680LT. NewBlot Nitro is intended for nitrocellulose membranes only, and is ideal for removal of secondary antibody.