Learn More
LICORbio NewBlot™ PVDF Stripping Buffer for PVDF Membranes (100 mL)
Catalog No. NC9804777
Encompass_Preferred
NewBlot PVDF stripping buffer is specially formulated for IRDye secondary antibodies including IRDye 800CW, IRDye 680RD, and IRDye 680LT. NewBlot PVDF is intended for PVDF membranes only and is ideal for removal of secondary antibody.
Catalog No. NC9804777 Supplier LICORbio Supplier No. 92840032
NewBlot PVDF stripping buffer is specially formulated for IRDye secondary antibodies including IRDye 800CW, IRDye 680RD, and IRDye 680LT. NewBlot PVDF is intended for PVDF membranes only and is ideal for removal of secondary antibody.