Learn More
American Research Products Inc New Delhi metallo beta lactama
Catalog No. 5000329581
Encompass
New Delhi metallo-beta-lactamase-1 (NDM-1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL- 400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma urine samples infected by New Delhi super bacteria Method Sandwich
Catalog No. 50-003-29581 Supplier American Research Products Inc Supplier No. CSBE14924
New Delhi metallo-beta-lactamase-1 (NDM-1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL- 400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma urine samples infected by New Delhi super bacteria Method Sandwich