Learn More
Electron Microscopy Sciences EMS Tungsten Loop Filament New, Hitachi S-Type
SDP
Catalog No. 50949769
Encompass
Our Electron microscope Tungsten filaments have a rigid attachment of the filament wire to the post with no possiblity of waving and a minimum tip radius for a coherent beam. The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. Inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament.
Use: Hitachi S-Type
Catalog No. 50-949-769 Supplier Electron Microscopy Sciences Supplier No. 81022
Our Electron microscope Tungsten filaments have a rigid attachment of the filament wire to the post with no possiblity of waving and a minimum tip radius for a coherent beam. The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. Inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament.
Use: Hitachi S-Type