Learn More
Electron Microscopy Sciences EMS Tungsten Loop Filament New, AEI, Cambridge S Series, Novascan, Semco, and Nanolab
SDP
Catalog No. 5029076
Encompass_Preferred
Our Electron microscope Tungsten filaments have a rigid attachment of the filament wire to the post with no possiblity of waving and a minimum tip radius for a coherent beam. The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. Inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament.
Use: AEI, Cambridge S Series, Novascan, Semco, and Nanolab
Catalog No. 50-290-76 Supplier Electron Microscopy Sciences Supplier No. 81020
Our Electron microscope Tungsten filaments have a rigid attachment of the filament wire to the post with no possiblity of waving and a minimum tip radius for a coherent beam. The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. Inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament.
Use: AEI, Cambridge S Series, Novascan, Semco, and Nanolab