Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Fuchsin [for Biochemical Research], TCI America™
SDP

Catalog No. N08735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08735G Supplier TCI America Supplier No. N08735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3248-91-7
Molecular Formula C22H24ClN3
Molecular Weight (g/mol) 365.91
MDL Number MFCD00012567
InChI Key ORDGVBRMUJCHEO-UHFFFAOYSA-M
Synonym new fuchsin, magenta iii, basic violet 2, neofuchsine, new fuchsine, new magenta, fuchsine sbp, c.i. basic violet 2, c.i. basic violet 2, monohydrochloride, new-fuchsin
PubChem CID 14454445
ChEBI CHEBI:87671
IUPAC Name tris(4-amino-3-methylphenyl)methylium chloride
SMILES [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1

Specifications

Quantity 5 g
Formula Weight 365.91
Physical Form Crystalline Powder
Chemical Name or Material New Fuchsin [for Biochemical Research]
EINECSNumber (5)-1970
RTECSNumber CX9825000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.