Learn More
AdipoGen New Indocyanine Green
SDP
Catalog No. 502560358
Encompass_Preferred
Chemical. CAS 172616-80-7. Formula C46H50ClN2NaO6S2. MW 849.47. IR-820 is a near infrared dye NIR, which has a broad absorption band at 690nm in aqueous solution, with high resistance to photodegradation and photocleavage. IR-820 is an excellent probe for imaging tumors, as it can be detected in skin cells, chitosan quaternary ammonium, and subcutaneous tumors using fluorescence imaging. IR-820 is an infrared blood pool contrast agent to detect and quantify diseased tissue in live animals. IR-820 has maximal excitation and emission wavelengths of 710 and 820nm, respectively. IR-820 emission is significantly improved in vivo on serum binding to albumin, and elimination occurs predominantly via the gastrointestinal tract. IR820 has optical and thermal generation properties similar to those of indocyanine green ICG but with improved in vitro and in vivo stability.
Catalog No. 50-256-0358 Supplier AdipoGen Supplier No. CDXN0516G001
Chemical. CAS 172616-80-7. Formula C46H50ClN2NaO6S2. MW 849.47. IR-820 is a near infrared dye NIR, which has a broad absorption band at 690nm in aqueous solution, with high resistance to photodegradation and photocleavage. IR-820 is an excellent probe for imaging tumors, as it can be detected in skin cells, chitosan quaternary ammonium, and subcutaneous tumors using fluorescence imaging. IR-820 is an infrared blood pool contrast agent to detect and quantify diseased tissue in live animals. IR-820 has maximal excitation and emission wavelengths of 710 and 820nm, respectively. IR-820 emission is significantly improved in vivo on serum binding to albumin, and elimination occurs predominantly via the gastrointestinal tract. IR820 has optical and thermal generation properties similar to those of indocyanine green ICG but with improved in vitro and in vivo stability.