Learn More
Waters Corp Waters Injector Upgrade Kit, Second Generation, for 2695 Alliance HPLC Systems
Catalog No. 50792804 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit upgrades older Alliance HPLC Systems to incorporate a second-generation injector design. The new design features fewer moving parts, enhancing reliability and robustness. It is compatible with 2695 and 2695D Separations Modules.
  • Enables incorporation of second-generation injector design.
  • New injector design features fewer moving parts.
  • Enhances instrument reliability and robustness.
  • Can be retrofitted into any 2695 or 2695D Separations Module.
  • Designed for 2695 separations module and 2695D dissolution separations module.
  • Recommended for use with the new second-generation injector design.
  • Facilitates faster and easier column connections when used with appropriate tubing kits.
Catalog No. 50-792-804 Supplier Waters Corp Supplier No. 700002790
This kit upgrades older Alliance HPLC Systems to incorporate a second-generation injector design. The new design features fewer moving parts, enhancing reliability and robustness. It is compatible with 2695 and 2695D Separations Modules.
  • Enables incorporation of second-generation injector design.
  • New injector design features fewer moving parts.
  • Enhances instrument reliability and robustness.
  • Can be retrofitted into any 2695 or 2695D Separations Module.
  • Designed for 2695 separations module and 2695D dissolution separations module.
  • Recommended for use with the new second-generation injector design.
  • Facilitates faster and easier column connections when used with appropriate tubing kits.