Learn More
Waters Corp New Method Scouting Kit, 4.6 mm ID, 100 mm Length, 3.5 μm Particle Size, 5 pk
Catalog No. 502391379 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides five 4.6 mm x 100 mm, 3.5 μm reversed-phase columns for general purpose method development and scouting. It streamlines HPLC method development for diverse chemistries, offering orthogonal selectivity profiles for quicker screening and confident results. Manufactured to high quality standards, these columns ensure reproducibility, high resolution, and extended life, suitable for pharmaceuticals, biopharmaceuticals, environmental testing, and food analysis.
  • Includes five 4.6 mm x 100 mm, 3.5 μm columns.
  • Offers orthogonal selectivity profiles.
  • Ensures reproducibility, high resolution, and extended column life.
  • Streamlines HPLC method development.
  • Supports diverse compound chemistries.
  • Suitable for pharmaceutical, environmental, and food analysis.
Catalog No. 50-239-1379 Supplier Waters Corp Supplier No. 176003359
Add to Cart
Add to Cart
Request a Quote
This kit provides five 4.6 mm x 100 mm, 3.5 μm reversed-phase columns for general purpose method development and scouting. It streamlines HPLC method development for diverse chemistries, offering orthogonal selectivity profiles for quicker screening and confident results. Manufactured to high quality standards, these columns ensure reproducibility, high resolution, and extended life, suitable for pharmaceuticals, biopharmaceuticals, environmental testing, and food analysis.
  • Includes five 4.6 mm x 100 mm, 3.5 μm columns.
  • Offers orthogonal selectivity profiles.
  • Ensures reproducibility, high resolution, and extended column life.
  • Streamlines HPLC method development.
  • Supports diverse compound chemistries.
  • Suitable for pharmaceutical, environmental, and food analysis.