Learn More
Volu Sol New Methylene Blue, 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. NC9901594 Shop All Volu Sol Products
Encompass_Preferred
Used as a blood and reticulocyte staining solution.  When mixed with whole blood, the RNA and DNA stain deep blue
Catalog No. NC9901594 Supplier Volu Sol Supplier No. VMB016
Used as a blood and reticulocyte staining solution.  When mixed with whole blood, the RNA and DNA stain deep blue