Learn More
New Methylene Blue N Zinc Chloride Double Salt, Sigma-Aldrich
Shop All MilliporeSigma Supelco ProductsDescription
New Methylene Blue N zinc chloride double salt also known as methylene blue N is an azo dye. It can bind, neutralize and crosslink with RNA. This causes co-precipitation of ribosomal and residual RNA with the mitochondria and ferritin masses. These events make the living, young erythrocytes as microscopically visible, dark-blue clusters and filaments, referred to as reticulum. New Methylene Blue N zinc chloride double salt helps in counting of reticulocytes (erythrocytes containing RNA).
Specifications
Specifications
| Chemical Name or Material | Methylene blue N |
| CAS | 6586-05-6 |
| Color | Blue |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 416.05 g/mol |
| Percent Purity | 90% |
| Grade | Not Available |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.