Learn More
Crystal Chem Inc New Mouse Total GIP ELISA Kit, 96-tests, Uses 10uL per sample
SDP
Supplier:  Crystal Chem Inc 81527
Encompass
This NEW and update Mouse GIP ELISA kit uses only a 10 µL sample and has a wide dynamic range to quantify levels of mouse GIP in plasma and cell culture.Highlights: Kits use a sample volume of only 10 µLWide dynamic range of 2.5 - 600 pM. High sensitivity with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7906