Learn More
MSC New Pig Traffic Mat, Poly-Backed, 36x100 ft., 1 roll
Catalog No. 5000674230
Encompass
This absorbent mat is New Pig's most durable for areas with heavy foot, cart, or forklift traffic. Its poly backing prevents liquids from reaching the floor, making it suitable for everyday spills and machine overspray in walkways and aisleways.
  • Stands up to heavy foot, industrial vehicle, and forklift traffic
  • Cuts easily with scissors or utility knife without ripping, tearing, or fraying
  • Rapidly absorbs oils, coolants, solvents, and water for faster cleanup
  • Can be wrung out and incinerated after use to reduce waste
  • Constructed from polypropylene for durability
  • Flame resistant, designed to melt and self-extinguish
  • Measures 36 in. W x 100 ft. L with a mediumweight design
  • Absorbs up to 13 gal. per roll
Catalog No. 50-006-74230 Supplier MSC Supplier No. 76151349
This absorbent mat is New Pig's most durable for areas with heavy foot, cart, or forklift traffic. Its poly backing prevents liquids from reaching the floor, making it suitable for everyday spills and machine overspray in walkways and aisleways.
  • Stands up to heavy foot, industrial vehicle, and forklift traffic
  • Cuts easily with scissors or utility knife without ripping, tearing, or fraying
  • Rapidly absorbs oils, coolants, solvents, and water for faster cleanup
  • Can be wrung out and incinerated after use to reduce waste
  • Constructed from polypropylene for durability
  • Flame resistant, designed to melt and self-extinguish
  • Measures 36 in. W x 100 ft. L with a mediumweight design
  • Absorbs up to 13 gal. per roll