Learn More
STEMCELL Technologies EasySep™ Human Pan-Granulocyte Isolation Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC0949888
Encompass
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-granulocytes and magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
Catalog No. NC0949888 Supplier STEMCELL Technologies Supplier No. 19259
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-granulocytes and magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.