Learn More
LICORbio NewBlot™ IR Stripping Buffers for NIR Western Blots (100 mL)
Catalog No. NC0804471
Encompass_Preferred
NewBlot IR is a robust formulation optimized for near-infrared (NIR) Western blots that works with both nitrocellulose and PVDF membranes. This stripping buffer is a good place to start if you are new to stripping and reprobing NIR Western blots. NewBlot IR works well for removal of all IRDye secondary antibodies, and may be suitable for removal of other dye-conjugated secondary antibodies. NewBlot IR does not require hazardous shipping, unlike many other stripping buffers. Note: NewBlot IR Stripping Buffer is not recommended for loading amounts over 30 µg.
Catalog No. NC0804471 Supplier LICORbio Supplier No. 92840028
NewBlot IR is a robust formulation optimized for near-infrared (NIR) Western blots that works with both nitrocellulose and PVDF membranes. This stripping buffer is a good place to start if you are new to stripping and reprobing NIR Western blots. NewBlot IR works well for removal of all IRDye secondary antibodies, and may be suitable for removal of other dye-conjugated secondary antibodies. NewBlot IR does not require hazardous shipping, unlike many other stripping buffers. Note: NewBlot IR Stripping Buffer is not recommended for loading amounts over 30 µg.