Learn More
Enterprise Technology Solutions Newell Brands China Maker,Peel-Off,f/Porous&Non-Porous Surfaces,Fade&Water-Resist,Wax,1 Dozen,Red
SDP
Catalog No. 5000824899
Encompass_Preferred
Write clear, legible marks on a variety of materials including porcelain, glass, metal and plastic with Sharpie Peel-Off China Markers. These versatile grease markers write smoothly on most porous and non-porous surfaces. Even better, there's no sharpening required - simply tear the string for more pigment. Each wax pencil has pigment that's both fade-resistant and water-resistant for marks that can withstand abuse. Plus, cleanup is simple, erasing easily with a damp cloth.
Catalog No. 50-008-24899 Supplier Enterprise Technology Solutions Supplier No. SAN2229138
Write clear, legible marks on a variety of materials including porcelain, glass, metal and plastic with Sharpie Peel-Off China Markers. These versatile grease markers write smoothly on most porous and non-porous surfaces. Even better, there's no sharpening required - simply tear the string for more pigment. Each wax pencil has pigment that's both fade-resistant and water-resistant for marks that can withstand abuse. Plus, cleanup is simple, erasing easily with a damp cloth.