Learn More
Enterprise Technology Solutions Newell Brands Paint Markers, Oil Base, Extra Fine, 12/BX,2/BD, White
SDP
Catalog No. 5000826679
Encompass_Preferred
Valve-action paint marker delivers an opaque and glossy effect on light or dark surfaces. Permanent oil-based opaque paint marks on virtually any surface including metal, pottery, wood, rubber, glass, plastic, stone, and more. Xylene-free paint marker contains fade-resistant, water-resistant and abrasion-resistant paint. Easily overlay colors and other mediums with this certified AP nontoxic.
Catalog No. 50-008-26679 Supplier Enterprise Technology Solutions Supplier No. SAN35531BD
Valve-action paint marker delivers an opaque and glossy effect on light or dark surfaces. Permanent oil-based opaque paint marks on virtually any surface including metal, pottery, wood, rubber, glass, plastic, stone, and more. Xylene-free paint marker contains fade-resistant, water-resistant and abrasion-resistant paint. Easily overlay colors and other mediums with this certified AP nontoxic.