Learn More
Enterprise Technology Solutions Newell Brands Paint Markers, Oil Base, Extra Fine, Metallic Gold
SDP
Catalog No. 5000825118
Encompass_Preferred
Valve-action paint marker delivers an opaque and glossy effect on light or dark surfaces. Permanent oil-based opaque paint marks on virtually any surface including metal, pottery, wood, rubber, glass, plastic, stone, and more. Xylene-free paint marker contains fade-resistant, water-resistant and abrasion-resistant paint. Easily overlay colors and other mediums with this certified AP nontoxic.
Catalog No. 50-008-25118 Supplier Enterprise Technology Solutions Supplier No. SAN35532
Valve-action paint marker delivers an opaque and glossy effect on light or dark surfaces. Permanent oil-based opaque paint marks on virtually any surface including metal, pottery, wood, rubber, glass, plastic, stone, and more. Xylene-free paint marker contains fade-resistant, water-resistant and abrasion-resistant paint. Easily overlay colors and other mediums with this certified AP nontoxic.