Learn More
Enterprise Technology Solutions Newell Brands Sharpie Industrial Marker, Fine Point, 36/PK, Black Ink
SDP
Catalog No. 5000824704
Encompass_Preferred
Industrial Marker with fine point is ideal for industrial, laboratory and specialty use. Permanent ink remains permanent on metal, foil, glass, photographic film, plastic and most hard-to-mark surfaces, even in extreme heat and steam. It is designed to withstand extreme heat - up to 500 degrees Fahrenheit (260 Celsius). Marker is water-resistant and smear-proof.
Catalog No. 50-008-24704 Supplier Enterprise Technology Solutions Supplier No. SAN2003898
Industrial Marker with fine point is ideal for industrial, laboratory and specialty use. Permanent ink remains permanent on metal, foil, glass, photographic film, plastic and most hard-to-mark surfaces, even in extreme heat and steam. It is designed to withstand extreme heat - up to 500 degrees Fahrenheit (260 Celsius). Marker is water-resistant and smear-proof.