Learn More
Enterprise Technology Solutions Newell Brands Sharpie Paint Marker, Oil-Based, Fine, 5/PK, AST
SDP
Catalog No. 5000825131
Encompass_Preferred
Create eye-catching opaque and glossy text and designs with the Sharpie Oil-Based Paint Marker. It works well on virtually any surface, including metal, pottery, wood, rubber, glass, plastic, stone and more. Permanent oil-based paint is released by a valve for simple, fluid control. Nontoxic formula is safe for use by children and adults so you can work on art projects with your little ones. Ink is opaque and glossy, even on dark surfaces. Quick-drying is also fade-resistant, water-resistant, abrasion-resistant and xylene-free. Easily overlay colors and other mediums. Fine point allows detailed drawing and coloring. Assorted colors include black, white, blue, yellow and red.
Catalog No. 50-008-25131 Supplier Enterprise Technology Solutions Supplier No. SAN37371PP
Create eye-catching opaque and glossy text and designs with the Sharpie Oil-Based Paint Marker. It works well on virtually any surface, including metal, pottery, wood, rubber, glass, plastic, stone and more. Permanent oil-based paint is released by a valve for simple, fluid control. Nontoxic formula is safe for use by children and adults so you can work on art projects with your little ones. Ink is opaque and glossy, even on dark surfaces. Quick-drying is also fade-resistant, water-resistant, abrasion-resistant and xylene-free. Easily overlay colors and other mediums. Fine point allows detailed drawing and coloring. Assorted colors include black, white, blue, yellow and red.