Learn More
Enterprise Technology Solutions Newell Brands Sharpie Paint Marker, Oil-Based, Medium Point, Blue
SDP
Catalog No. 5000825180
Encompass_Preferred
Sharpie oil-based paint marker is perfect for arts and crafts. Marker applies smooth and vivid colors. Ink is opaque and glossy, even on dark surfaces. Marker makes permanent marks on most surfaces including glass, metal, plastic, rubber, stone, wood, pottery and more. Certified AP nontoxic, quick-drying ink is weather-resistant, fade-resistant, acid-free and xylene-free.
Catalog No. 50-008-25180 Supplier Enterprise Technology Solutions Supplier No. SAN35551
Sharpie oil-based paint marker is perfect for arts and crafts. Marker applies smooth and vivid colors. Ink is opaque and glossy, even on dark surfaces. Marker makes permanent marks on most surfaces including glass, metal, plastic, rubber, stone, wood, pottery and more. Certified AP nontoxic, quick-drying ink is weather-resistant, fade-resistant, acid-free and xylene-free.