Learn More
Enterprise Technology Solutions Newell Brands T.E.C. Permeant Marker, Fine Point, Black
SDP
Catalog No. 5000825309
Encompass_Preferred
Sharpie T.E.C. Marker marks on most surfaces and meets requirements for trace element certified ink for the nuclear energy, aerospace, electronics, aeronautic and ship building industries. Chloride and other halogens cause stress corrosion on metals and should not be used on stainless steel or titanium. Fine-tip permanent marker is trace element certified. Design also features a cap to protect the ink and a pocket clip for attaching the marker to notebooks, pockets and more.
Catalog No. 50-008-25309 Supplier Enterprise Technology Solutions Supplier No. SAN13401
Sharpie T.E.C. Marker marks on most surfaces and meets requirements for trace element certified ink for the nuclear energy, aerospace, electronics, aeronautic and ship building industries. Chloride and other halogens cause stress corrosion on metals and should not be used on stainless steel or titanium. Fine-tip permanent marker is trace element certified. Design also features a cap to protect the ink and a pocket clip for attaching the marker to notebooks, pockets and more.