Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Newir Birch O-Pctm C0103

Catalog No. 914797100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-479-7100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-479-7100MG Supplier Sigma Aldrich Supplier No. 914797100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Birch O-PC(TM) C0103 is an organic photocatalyst developed in the Miyake lab for the photocatalytic Birch reduction driven by visible light. The hallmark high excited state redox potential of this first of its kind catalyst class reduces benzene and other arenes to produce functionally useful 1,4-cyclohexadienes. This organic photocatalyst accesses both classical and non-classical Birch reduction reactivity in addition to demonstrating hydrodechlorination of aromatics. Employing mild benchtop conditions, this photocatalytic approach eliminates challenges associated with traditional Birch reduction such as the use of alkali metals in liquid ammonia.Sold in collaboration with New Iridiu mLLC. For orders greater than 25g, please contact New Iridium at chern@newiridium.com or visit https://www.newiridium.com.

Specifications

CAS 384184-42-1
Linear Formula C50H63ClF3N5O12
Quantity 100 mg
Synonym AVP ligand; Ethyl (S)-2-((S)-1-((S)-2-((S)-2-aminopropanamido)-3,3-dimethylbutanoyl)pyrrolidine-2-carboxamido)-3-(4-fluorophenyl)propanoate; IAP E3 ligase lead for protein degrader research; SNIPER building block
Molecular Weight (g/mol) 492.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.