Learn More
Let'S Gel Inc NEWLIFE ECO-PRO 36X60 BLACK EA
SDP
Catalog No. 50489579
Encompass_Preferred
Anti-Fatigue Floor Mat, Commercial Floor Mat, Ergonomic Mat, Safety Mat, Newlife Eco-Pro 36 in.x60 in. Black, 100% Polyurethane Foam
Catalog No. 50-489-579 Supplier Let'S Gel Inc Supplier No. 1040136601
Anti-Fatigue Floor Mat, Commercial Floor Mat, Ergonomic Mat, Safety Mat, Newlife Eco-Pro 36 in.x60 in. Black, 100% Polyurethane Foam