Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning All About Animals Flip Chart
SCI_ED

Catalog No. S02533 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02533 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02533 Supplier NewPath Learning Supplier No. 346303
Add to Cart
Add to Cart

Includes

Ten charts: The Animal Kingdom, Vertebrates, Invertebrates, Animal Ranges, Physical Adaptations, Behavioral Adaptations, Animal Life Cycles, Animal Locomotion, Animal Habitats, Animal Tracks and Signs

Provides comprehensive coverage of key standards-based curriculum in an illustrated format that is visually appealing, engaging and easy to use in the entire classroom, learning centers or by students working independently.

  • Each Curriculum Mastery™ Flip Chart is mounted on a sturdy easel and features
  • 10 Double-sided, laminated 12 x 18 in. charts
  • Side one features a colorful, graphic overview of the topic
  • Side two serves as a write-on/wipe-off activity chart featuring questions, labeling exercises, vocabulary review & more
  • Activity Guide featuring black-line copy-masters & exercises

Specifications

Product Type All About Animals Flip Chart Set
Includes Ten charts: the animal kingdom, vertebrates, invertebrates, animal ranges, physical adaptations, behavioral adaptations, animal life cycles, animal locomotion, animal habitats, animal tracks and signs
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Intermediate - Middle School, High School
Grade 6 to 12
Subject Science
Material Laminated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.