Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

All About Plants Flip Chart Set
SCI_ED

Catalog No. S02534 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02534 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02534 Supplier NewPath Learning Supplier No. 346304
Add to Cart
Add to Cart

Includes

Ten charts: The Plant Kingdom, Plant Cells & Photosynthesis, Pollination & Germination, Gymnosperm vs. Angiosperm, Structure & Function of Plants, Adaptations of Plants, Plant Reproduction, Carnivorous Plants, Plants/Trees of North America, Flowering vs. Non-Flowering

Illustrated format is visually appealing, engaging and easy to use.

Provides comprehensive coverage of key standards-based curriculum in a simple, illustrated format. Can be used to teach the entire classroom, learning centers or students working independently.

  • Each Curriculum Mastery™ Flip Chart is mounted on a sturdy easel
  • Includes 10 double-sided, laminated charts
  • Dimensions: 12 x 18 in. (30.5 x 45.7cm)
  • Side one features a colorful, graphic overview of the topic
  • Side two serves as a write-on/wipe-off activity chart featuring questions, labeling exercises, vocabulary review and more
  • Activity Guide features black-line copy-masters and exercises

Specifications

Product Type All About Plants Flip Chart Set
Includes Ten charts: the plant kingdom, plant cells and photosynthesis, pollination and germination, gymnosperm vs. angiosperm, structure and function of plants, adaptations of plants, plant reproduction, carnivorous plants, plants/trees of north America, flowering vs. non-flowering
For Use With (Application) Can be used to teach the entire classroom, learning centers or students working independently
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Elementary, Intermediate - Middle School, High School
Grade 5 to 12
Subject Science
Material Laminated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.