Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Curriculum Mastery™ Flip Charts: Science
SCI_ED

Catalog No. S93941 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
1
2
3
4
5
6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S93941 3
S93939 1
S93942 4
S93940 2
S93943 5
S93944 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S93941 Supplier NewPath Learning Supplier No. 343001
Add to Cart
Add to Cart

Includes

Activity guide with black-line copy-masters, review questions, vocabulary terms and suggested activities

Presents comprehensive review of science standards.

Curriculum Mastery Flip Charts provide a hands-on review of standards-based curriculum skills. Each developed to address the standards for its specific grade.

  • 10 double-sided, laminated charts
  • Side 1 shows a colorful, graphic overview of the topic; Side 2 serves as a write-on/wipe-off activity chart featuring questions, labeling exercises, vocabulary review and more
  • Charts are mounted on sturdy easels for years of use
  • Chart Dimensions (L × W): 18 × 12 in. (45.7 × 30.5cm)

Topics include
Grade 1 (S93939):

  • Living and Nonliving Things; Where Do Animals Live?; Life Cycles; Food Chains; My Senses; Human Body; Weather; The Seasons; The Sky; and Land, Water and Air
Grade 2 (S93940):
  • All About Plants; All About Animals; Land Habitats; Water Habitats; How Living Things Change and Grow; Mammals and Birds; Reptiles, Amphibians and Fish; The Sun and Earth; Moon, Stars, and Planets; and Fossils and Dinosaurs
Grade 3 (S93941):
  • Main Parts of Plants; How Do Plants Grow?; Grouping of Plants; Grouping of Animals; Relationships Among Living Things; Rocks, Minerals and Soil; Changes on Earth; Matter and Its Properties; Work and Machines; and the Solar System
Grade 4 (S93942):
  • Cells; Invertebrates; Vertebrates; Animal Growth and Reproduction; Food Chains and Food Webs; Rocks and Minerals; Earth: Inside and Out; Weather and Climate; Force, Motion, and Energy; and Electricity and Magnetism
Grade 5 (S93943):
  • The Six Kingdoms of Life; Roots, Stems, and Leaves; Flowers and Seeds; Cycles of Life and Biomes; Landforms, Rocks, and Soil; Earth's Freshwater and Atmosphere; Elements, Mixtures, and Compounds; Acids and Bases; Newton's Laws of Motion; and Sound and Light
Grade 6 (S93944):
  • Diversity of Life; Cells: The Basic Unit of Life; Genetics; Introduction to Plants; Introduction to Animals; Ecosystems; Volcanoes; The Sun-Earth-Moon System; Mixtures, Solutions, and Compounds; and Electromagnetism

Specifications

Grade Level Elementary
Grade 3
For Use With (Application) Labeling exercises, vocabulary review and more
Product Type Science Skills Flip Chart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.