Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gene Technology Poster
SCI_ED

Catalog No. S02537 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02537 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02537 Supplier NewPath Learning Supplier No. 346107
Add to Cart
Add to Cart

Features detailed information and illustrations on DNA fingerprinting, genetic engineering, vaccine production and Polymerase Chain Reaction (PCR).

  • Colorful, visually appealing
  • Features detailed illustrations to facilitate comprehension of key, standards-based concepts
  • Dimensions: 23 x 35 in. (58.4 x 88.9cm)
  • Laminated for years of use

Specifications

Product Type Gene Technology Poster
Includes Detailed information and illustrations on DNA fingerprinting, genetic engineering, vaccine production and polymerase chain reaction
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Elementary, Intermediate - Middle School, High School
Grade 5 to 12
Subject Science
Material Laminated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.