Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning I Love Math Hands-On Program
SCI_ED

Catalog No. S01484 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
1
2
3
4
5 to 12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S01484 5 to 12
S01480 1
S01483 4
S01482 3
S01481 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S01484 Supplier NewPath Learning Supplier No. 135010
Add to Cart
Add to Cart

Complete coverage of NCTM Standards covering Grades 1 to 5.

Complete coverage of 15 concepts: Whole Numbers, Patterns and Algebra, Mass, Addition and Subtraction, Data, Time, Multiplication and Division, Length, Three-Dimensional Space, Chance, Volume and Capacity, Position

  • Developed by an experienced educator and classroom-tested for more than a decade
  • Wealth of challenging, mathematical exercises focused on leveled, standards-based topics
  • Fun-filled means of motivating students to think more deeply, investigate, explain, and understand problem-solving strategies
  • Concepts divided into two sections: one per semester
  • Each section is two to three pages in length and followed by an assessment for immediate and continuous feedback

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 5 to 12
Format E-books/Workbooks
Product Type I Love Hands-on Math Reproducible Workbook
For Use With (Application) Fun filled means of motivating students to think more deeply, investigate, explain and understand problem-solving strategies
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.