Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning Mastering Middle School Life Science Skills - Grade 6 to 8 Take-Home Game
SCI_ED

Catalog No. S95687 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95687 6 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95687 Supplier NewPath Learning Supplier No. 246807
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

25 Individually packaged games each covering a different topic and with a set of 30 standards-based review questions, double-sided game board, Teacher and Parent Planning and Resource Guide, presentation CD featuring interactive questions for PC, Mac or Interactive Whiteboard. Materials for up to four students.

Designed to stimulate outside review and parental involvement.

Provides comprehensive coverage of the following curriculum topics:

  • The World of Life Science; Diversity of Life; Cells: The Basic Units of Life; Cell Processes; Cell Reproduction; Genetics – Study of Heredity; Modern Genetics; The Evolution and Interaction of Living Things; Bacteria and Viruses; Protists and Fungi, Introduction to Plants; Plant Processes; Plant Reproduction; Introduction to Animals; Sponges, Cnidarians and Worms; Mollusks, Arthropods and Echinoderms; Fishes, Amphibians and Reptiles; Birds and Mammals; Bones, Muscles and Skin; The Digestive System and Nutrition; Circulation and Immunity; Respiration and Excretion; The Nervous System; The Endocrine System and Reproduction; Ecosystems; Food Chains and Food Webs

Order Info

Every game is individually packaged in a vinyl folder.

Specifications

Grade Level Intermediate - Middle School
Grade 6 to 8
For Use With (Application) Stimulate outside review and parental involvement
Includes 25 Individually packaged games each covering a different topic and with a set of 30 standards-based review questions, double-sided game board, Teacher and Parent Planning and Resource Guide, presentation CD featuring interactive questions for PC, Mac or Interactive Whiteboard
Description Life Science Skills - Grade 6 to 8 Take-Home Games
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.