Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Middle School NGSS - Earthquakes Student Learning Guide
SCI_ED

Catalog No. s04898 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04898 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04898 Supplier NewPath Learning Supplier No. 146824
Only null left
Add to Cart
Add to Cart

Full-color comprehensive coverage of key Next Generation Science Concepts for grades 6 - 10.

  • Self-directed readings
  • Easy-to-follow illustrated explanations
  • Guiding questions
  • Inquiry-based activities
  • Lab investigation
  • Key vocabulary review
  • Assessment review questions and post-test

Topics Covered

  • How an Earthquake Occurs
  • Types of Stress in Crustal Rock
  • Faults in the Earth's Crust
  • How Earth's Surface Changes
  • Seismic Waves
  • Measuring Earthquakes
  • The Richter Scale
  • Earthquake Destruction
  • Earthquake Safety
  • Vocabulary Review

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Product Type Earthquakes Learning Guide
For Use With (Application) Grades 6-10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.