Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Middle School NGSS - Light and Optics Student Learning Guide
SCI_ED

Catalog No. S04924 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04924 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04924 Supplier NewPath Learning™ Supplier No. 146929
Add to Cart
Add to Cart

Full-color comprehensive coverage of key Next Generation Science Concepts for grades 6 – 10.

  • Self-directed readings
  • Easy-to-follow illustrated explanations
  • Guiding questions
  • Inquiry-based activities
  • Lab investigation
  • Key vocabulary review
  • Assessment review questions and post-test
  • Topics Covered
  • Introduction to Light
  • The EM Spectrum
  • Transmission of Light
  • Light and Color
  • Interactions with Light
  • Reflections and Mirrors
  • Refraction and Lenses
  • Light and the Human Eye (Vision)
  • Light in Technology
  • Vocabulary Review

Specifications

Product Type Light and Optics Learning Guide
Format 49-page workbook
Grade Level Intermediate - Middle School, High School
Grade 6 to 12
Subject Physical Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.