Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Osmosis and Diffusion Flip Chart Set
SCI_ED

Catalog No. S05004 Shop All NewPath Learning Products
Change view
Click to view available options
Format:
Flip Chart Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S05004 Flip Chart Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05004 Supplier NewPath Learning™ Supplier No. 346723
Add to Cart
Add to Cart

Graphically Introduce and Review Key Curriculum Topics!

Introduce and review standards-based Science concepts. The set features 10 double-sided, laminated charts, each 12 in. x 18 in. and spiral-bound on a durable, free-standing easel –

  • Side 1 of each chart features a graphic overview of topic
  • Side 2 serves as write-on/wipe-off activity chart

Also includes Activity Guide featuring copy-masters and exercises.

Provides comprehensive coverage of the following topics –

  • Comparing Plant and Animal Cells
  • Cell Membrane and Cell Transport
  • Diffusion
  • Diffusion in the Lungs
  • Osmosis: The Diffusion of Water
  • Passive Transport
  • Active Transport
  • Osmosis in Plant Cells
  • Osmosis in Animal Cells
  • Vocabulary Review

Specifications

Format Flip Chart Set
Product Type Osmosis Flip Chart Set
For Use With (Application) Grades 6 to 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.