Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Six Kingdoms Flip Chart Set
SCI_ED

Catalog No. S04879 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04879 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04879 Supplier NewPath Learning™ Supplier No. 346729
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Graphically Introduce and Review Key Curriculum Topics!

Introduce and review standards-based Science concepts. The set features 10 double-sided, laminated charts, each 12 in. x 18 in. and spiral-bound on a durable, free-standing easel –

  • Side 1 of each chart features a graphic overview of topic
  • Side 2 serves as write-on/wipe-off activity chart

Also includes Activity Guide featuring copy-masters and exercises.

Provides comprehensive coverage of the following topics –

  • Classification
  • The Six Kingdoms
  • Archaebacteria and Eubacteria
  • Protista
  • Fungi
  • Plant Kingdom
  • Plants with Seeds
  • Animal Kingdom
  • Vertebrates and Invertebrates
  • Vocabulary Review

Specifications

Grade Level Intermediate - Middle School, High School
Grade 6 to 10
Product Type The Six Kingdoms Flip Chart Set
For Use With (Application) Grades 6 to 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.