Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Next Generation Science Standards Physical Science Vocabulary Builder Flash Card Set- Middle School
SCI_ED

Catalog No. S04935 Shop All NewPath Learning Products
Change view
Click to view available options
Format:
Flash Card Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S04935 Flash Card Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04935 Supplier NewPath Learning Supplier No. 446019
Add to Cart
Add to Cart

Includes

Teacher Resource Guide with suggested activities and trial subscription to Online Learning Program

Flash cards provide comprehensive coverage of key terms in disciplinary core ideas of Next Generation Science Standards and current state standards.

Vocabulary builder flash card sets can be used for conducting a variety of engaging activities and games which foster lasting understanding of key concepts.

  • Each card features term, pronounciation, graphic depiction on one side and definiton of term on other side
  • Colorfully illustrated cards printed on 10.1 x 14cm (4 x 5.5 in.) heavy stock
  • Used on bulletin boards or as pocket cards
  • For individual or small group review
  • Set has 200 flash cards

Specifications

Format Flash Card Set
Product Type Physical Science Vocabulary Builder Flash Card Set
For Use With (Application) Middle School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.