Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Next Generation Science Standards Science Vocabulary Builder Flash Card Sets
SCI_ED

Catalog No. S04932 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04932 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04932 Supplier NewPath Learning Supplier No. 443011
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Teacher Resource Guide with suggested activities and trial subscription to Online Learning Program

Flash cards provide comprehensive coverage of key terms in disciplinary core ideas of Next Generation Science Standards and current state standards.

Vocabulary builder flash card sets can be used for conducting a variety of engaging activities and games which foster lasting understanding of key concepts.

  • Each card features term, pronounciation, graphic depiction on one side and definiton of term on other side
  • Colorfully illustrated cards printed on 10.1 x 14cm (4 x 5.5 in.) heavy stock
  • Used on bulletin boards or as pocket cards
  • For individual or small group review
  • Eact set has 200 flash cards

Specifications

Grade Level Elementary
Grade 3 to 5
Product Type Vocabulary Builder Flash Card Set
Quantity 200 Cards
For Use With (Application) Vocabulary and reading skills
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.