Learn More
Description
Full-color, 36-page guide features self-directed readings, easy-to-follow illustrated explanations; guiding questions; inquiry-based activities; a lab investigation; key vocabulary review; and assessment review questions along with a post-test
Topics Covered
- Environment
- Living and Nonliving Things
- What is an Ecosystem?
- An Organism's Habitat and Niche
- Population and Community
- Parts of an Ecosystem
- How Does an Ecosystem Work?
- Kinds of Ecosystems
- Energy in an Ecosystem
- Producers, Consumers and Decomposers
- How Do Ecosystems Change?
- Environmental Changes and Effects on Organisms
Specifications
Specifications
| Product Type | Learning Guide |
| Format | 36-page workbook |
| For Use With (Application) | Enhance student comprehension on standards-based concepts on ecosystems |
| Certifications/Compliance | Academic Standard for California, Florida, National, New York, Texas |
| Quantity | 1 Ea. |
| Grade Level | Elementary |
| Grade | 3 to 5 |
| Subject | Life Science |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
