Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mastering Learning Middle School Earth Science Skills - Grade 6 to 8 Take-Home Game
SCI_ED

Catalog No. s95688 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95688 6 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95688 Supplier NewPath Learning Supplier No. 246808
Only null left
Add to Cart
Add to Cart

Includes

25 Individually packaged games each covering a different topic and with a set of 30 standards-based review questions, double-sided game board, Teacher and Parent Planning and Resource Guide, presentation CD featuring interactive questions for PC, Mac or Interactive Whiteboard. Materials for up to four students.

Designed to stimulate outside review and parental involvement.

Provides comprehensive coverage of following curriculum topics:
Introduction to Earth Science; Maps as Models of the Earth/Contour Models; Minerals, Rocks, Fossils; Earth's Energy Resources; Geologic Time; Plate Tectonics; Earthquakes; Volcanoes; Weathering of Rocks and Soil Formation; Earth's Fresh Water; Groundwater Resources; Agents of Erosion and Deposition; Exploring the Oceans/Oceanography; The Movement of Ocean Water; Earth's Atmosphere; Understanding Weather; Weather Patterns; Climate; Studying and Exploring Space, Stars, Galaxies and the Universe; The Sun-Earth-Moon System; Our Solar System; Our Impact on Earth

Order Info

Every game is individually packaged in a vinyl folder.

Specifications

Grade Level Intermediate - Middle School
Grade 6 to 8
For Use With (Application) Stimulate outside review and parental involvement
Includes 25 Individually packaged games each covering a different topic and with a set of 30 standards-based review questions, double-sided game board, Teacher and Parent Planning and Resource Guide, presentation CD featuring interactive questions for PC, Mac or Interactive Whiteboard
Description Earth Science Skills - Grade 6 to 8 Take-Home Game
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.