Learn More
Newcomer Supply NewSilane Adhesive Coated (Charged) Microscope Slides. Excellent for difficult and routine IHC procedures.
SDP
Catalog No. NC0498172
Encompass
The NewSilane adhesive coated slides are a very strong adhesive coated slide.  Use for routine Histo and/or IHC applications.  These adhesive coated slides are an excellent addition to any pathology lab that works with challenging tissue(s) or demanding procedures. Recommended use: difficult tissue combined with long incubations, extra procedural steps and extended antigen retrieval. Specifications: No word imprint on slide, Size: 75 x 25mm, 1mm thick, Edge Treatment: 90° Corner, Color: White, Unit: Approx. 72 slides/box, 1,440 slides/case
Catalog No. NC0498172 Supplier Newcomer Supply Supplier No. 5070
The NewSilane adhesive coated slides are a very strong adhesive coated slide.  Use for routine Histo and/or IHC applications.  These adhesive coated slides are an excellent addition to any pathology lab that works with challenging tissue(s) or demanding procedures. Recommended use: difficult tissue combined with long incubations, extra procedural steps and extended antigen retrieval. Specifications: No word imprint on slide, Size: 75 x 25mm, 1mm thick, Edge Treatment: 90° Corner, Color: White, Unit: Approx. 72 slides/box, 1,440 slides/case