Learn More
New England Biolabs, Inc. NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module – 96 reactions
SDP
Catalog No. 501959354
Encompass
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA. This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance
Catalog No. 50-195-9354 Supplier New England Biolabs, Inc. Supplier No. E6421L
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA. This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance