Learn More
New England Biolabs, Inc. NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina -24 rxns – 24 reactions
SDP
Catalog No. 501959353
Encompass
The NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA, followed by conversion to sequence-ready libraries using the Ultra II FS workflow. This unique workflow enables generation of the highest yields from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance. Please note that adaptors and primers are not included in the kit and are available separately.
Catalog No. 50-195-9353 Supplier New England Biolabs, Inc. Supplier No. E6420S
The NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA, followed by conversion to sequence-ready libraries using the Ultra II FS workflow. This unique workflow enables generation of the highest yields from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance. Please note that adaptors and primers are not included in the kit and are available separately.