Learn More
New England Biolabs, Inc. NEBNext Multiplex Small RNA Library Prep Kit for Illumina Set 2 – 96 reactions
SDP
Catalog No. 501959356
Encompass
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.
Catalog No. 50-195-9356 Supplier New England Biolabs, Inc. Supplier No. E7580L
Add to Cart
Add to Cart
Request a Quote
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.