Learn More
New England Biolabs, Inc. NEBNext Multiplex Oligos for Illumina (Dual Index Primers Set 2) – 96 reactions
SDP
Catalog No. 501527875
Encompass
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Catalog No. 50-152-7875 Supplier New England Biolabs, Inc. Supplier No. E7780S
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.