Learn More
New England Biolabs, Inc. NEBNext Multiplex Oligos for Illumina (96 Index Primers) – 384 reactions
SDP
Catalog No. 502024014
Encompass
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. The 96 8-base index primers included in this kit are pre-mixed with the universal primer and are packaged in a single-use 96-well plate with a pierceable foil seal. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Catalog No. 50-202-4014 Supplier New England Biolabs, Inc. Supplier No. E6609L
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. The 96 8-base index primers included in this kit are pre-mixed with the universal primer and are packaged in a single-use 96-well plate with a pierceable foil seal. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.