Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II DNA Library Prep Kit for Illumina® – 96 reactions
SDP
Catalog No. 501050632
Encompass
The Ultra II DNA Library Prep Kit for Illumina meets the challenge of constructing high quality libraries from ever-decreasing input quantities. The reagents for each step in the library preparation workflow have been reformulated to enable high yield preparation of high quality libraries from 500 picograms to 1 microgram of input DNA. This new generation of NEBNext reagents uses a fast, streamlined, automatable workflow and enables use of fewer PCR cycles while also improving GC coverage. The kit is also compatible with PCR-free workflows and is effective with challenging samples such as FFPE DNA.
  • Get more of what you need, with the highest library yields
  • Use to generate high quality libraries from a broad range of input amounts, from 500 pg to 1 µg
  • Prepare libraries from ALL your samples, including GC-rich DNA and FFPE DNA samples
  • Improve yields and quality for target enrichment applications
Catalog No. 50-105-0632 Supplier New England Biolabs, Inc. Supplier No. E7645L
The Ultra II DNA Library Prep Kit for Illumina meets the challenge of constructing high quality libraries from ever-decreasing input quantities. The reagents for each step in the library preparation workflow have been reformulated to enable high yield preparation of high quality libraries from 500 picograms to 1 microgram of input DNA. This new generation of NEBNext reagents uses a fast, streamlined, automatable workflow and enables use of fewer PCR cycles while also improving GC coverage. The kit is also compatible with PCR-free workflows and is effective with challenging samples such as FFPE DNA.
  • Get more of what you need, with the highest library yields
  • Use to generate high quality libraries from a broad range of input amounts, from 500 pg to 1 µg
  • Prepare libraries from ALL your samples, including GC-rich DNA and FFPE DNA samples
  • Improve yields and quality for target enrichment applications