Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II Q5® Master Mix – 250 reactions
SDP
Catalog No. 501050634
Encompass
The NEBNext Ultra II Q5 Master Mix is the most recent formulation of Q5 DNA polymerase that has been optimized for robust, high-fidelity amplification of next-generation sequencing (NGS) libraries. This new formulation further improves the uniformity of amplification of libraries, including superior performance with GC-rich regions. The polymerase component of the master mix, Q5 High-Fidelity DNA Polymerase, is a novel thermostable DNA polymerase that possesses 3´- 5´ exonuclease activity, and is fused to a processivity-enhancing Sso7d domain. Q5 also has the highest fidelity available (> 280-fold higher than that of Taq DNA Polymerase), resulting in ultra-low error rates. The NEBNext Ultra II Q5 Master Mix is an aptamer-based hot start formulation that allows simple room temperature reaction set up. The convenient 2X master mix format contains dNTPs, Mg++ and a proprietary buffer, and requires only the addition of primers and DNA template for robust amplification.
Catalog No. 50-105-0634 Supplier New England Biolabs, Inc. Supplier No. M0544L
The NEBNext Ultra II Q5 Master Mix is the most recent formulation of Q5 DNA polymerase that has been optimized for robust, high-fidelity amplification of next-generation sequencing (NGS) libraries. This new formulation further improves the uniformity of amplification of libraries, including superior performance with GC-rich regions. The polymerase component of the master mix, Q5 High-Fidelity DNA Polymerase, is a novel thermostable DNA polymerase that possesses 3´- 5´ exonuclease activity, and is fused to a processivity-enhancing Sso7d domain. Q5 also has the highest fidelity available (> 280-fold higher than that of Taq DNA Polymerase), resulting in ultra-low error rates. The NEBNext Ultra II Q5 Master Mix is an aptamer-based hot start formulation that allows simple room temperature reaction set up. The convenient 2X master mix format contains dNTPs, Mg++ and a proprietary buffer, and requires only the addition of primers and DNA template for robust amplification.