Learn More
New England Biolabs, Inc. NEBNext Ultra™ Ligation Module – 96 reactions
SDP
Catalog No. 501526365
Encompass
This module is part of the original Ultra workflow, and is optimized for use with the NEBNext Ultra End Repair/dA-Tailing Module, for Illumina-compatible library construction. The original Ultra workflow enables library preparation from 5 ng – 1 µg of input DNA in a streamlined protocol
Catalog No. 50-152-6365 Supplier New England Biolabs, Inc. Supplier No. E7445L
This module is part of the original Ultra workflow, and is optimized for use with the NEBNext Ultra End Repair/dA-Tailing Module, for Illumina-compatible library construction. The original Ultra workflow enables library preparation from 5 ng – 1 µg of input DNA in a streamlined protocol