Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II Directional RNA Library Prep Kit for Illumina® – 96 reactions
SDP
Catalog No. 501526372
Encompass
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the “dUTP” method for strand-specificity, with proven superiority for this application.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE
Catalog No. 50-152-6372 Supplier New England Biolabs, Inc. Supplier No. E7760L
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the “dUTP” method for strand-specificity, with proven superiority for this application.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE