Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II FS DNA Library Prep Kit for Illumina® – 24 reactions
SDP
Catalog No. 501527870
Encompass
The NEBNext Ultra II FS DNA Library Prep Kit for Illumina provides a fragmentation system: a fast and reliable solution for DNA fragmentation and library construction. A new DNA fragmentation reagent is combined with end repair and dA-tailing reagents, allowing these steps to be performed in the same tube, with no clean-ups or sample loss. The same fragmentation protocol is used for any input amount (100 pg–500 ng), or GC content. The kit also includes the NEBNext Ultra II Ligation Master Mix for adaptor ligation, and the NEBNext Ultra II Q5 Master Mix for uniform, high-fidelity library amplification.
  • Perform fragmentation, end repair and dA-tailing with a single enzyme mix
  • Experience reliable fragmentation with a single protocol, regardless of input amount or GC content
  • Prepare high quality libraries from a wide range of input amounts: 100 pg-500 ng
  • Use with DNA in standard buffers (TE, Tris-HCl) and water
Catalog No. 50-152-7870 Supplier New England Biolabs, Inc. Supplier No. E7805S
The NEBNext Ultra II FS DNA Library Prep Kit for Illumina provides a fragmentation system: a fast and reliable solution for DNA fragmentation and library construction. A new DNA fragmentation reagent is combined with end repair and dA-tailing reagents, allowing these steps to be performed in the same tube, with no clean-ups or sample loss. The same fragmentation protocol is used for any input amount (100 pg–500 ng), or GC content. The kit also includes the NEBNext Ultra II Ligation Master Mix for adaptor ligation, and the NEBNext Ultra II Q5 Master Mix for uniform, high-fidelity library amplification.
  • Perform fragmentation, end repair and dA-tailing with a single enzyme mix
  • Experience reliable fragmentation with a single protocol, regardless of input amount or GC content
  • Prepare high quality libraries from a wide range of input amounts: 100 pg-500 ng
  • Use with DNA in standard buffers (TE, Tris-HCl) and water