Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II RNA Library Prep Kit for Illumina® – 24 reactions
SDP
Catalog No. 501526376
Encompass
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1µg.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE
Catalog No. 50-152-6376 Supplier New England Biolabs, Inc. Supplier No. E7770S
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1µg.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE